Order Entry
Export
ContactUsLinkComponent
 
Anti-CD36 Mouse Monoclonal Antibody (CF405S) [clone: 185-1G2]
 :  Biotium
 
Anti-CD36 Mouse Monoclonal Antibody (CF405S) [clone: 185-1G2]
 :  Biotium

 

  • Catalog No:
  • BNC040169-500
  • BNC040169-100
  • Antigen symbol:
  • CD36
  • CD36
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone 185-1G2
  • Clone 185-1G2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 120949
  • 120949
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide