Order Entry
Export
ContactUsLinkComponent
Anti-ACTH Mouse Monoclonal Antibody (Purified) [clone: 57]
Anti-ACTH Mouse Monoclonal Antibody (Purified) [clone: 57]
Supplier:  Biotium
Certificates
Anti-ACTH Mouse Monoclonal Antibody (Purified) [clone: 57]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB0139-500
  • BNUB0139-100
  • Antigen symbol:
  • ACTH
  • ACTH
  • Antigen name:
  • Adrenocorticotropic hormone
  • Adrenocorticotropic hormone
  • Antigen synonyms:
  • alpha or beta or gamma Melanocyte Stimulating Hormone (MSH) or Melanotropin|beta-Endorphin|POC|beta or gamma Lipotropin (LPH)|CLIP|POMC|Adrenocorticotropin|Met Enkephalin
  • alpha or beta or gamma Melanocyte Stimulating Hormone (MSH) or Melanotropin|beta-Endorphin|POC|beta or gamma Lipotropin (LPH)|CLIP|POMC|Adrenocorticotropin|Met Enkephalin
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone 57
  • Clone 57
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 1897
  • 1897
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications