Order Entry
Export
ContactUsLinkComponent
Anti-H1 Mouse Monoclonal Antibody (CF640R) [clone: AE-4]
Anti-H1 Mouse Monoclonal Antibody (CF640R) [clone: AE-4]
Supplier:  Biotium
Certificates
Anti-H1 Mouse Monoclonal Antibody (CF640R) [clone: AE-4]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400096-500
  • BNC400096-100
  • Antigen symbol:
  • H1
  • H1
  • Antigen name:
  • Histone H1
  • Histone H1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone AE-4
  • Clone AE-4
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 226117 , 97358
  • 226117 , 97358
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications