Anti-CGA Mouse Monoclonal Antibody (Biotin) [clone: HCGa/53]
Supplier: Biotium
Certificates
About this item
This MAb reacts with a protein of ~13 kDa, identified as alpha subunit of HCG. HCG is a glycoprotein, which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH).
Type: Primary
Antigen: CGA
Clonality: Monoclonal
Clone: HCGa/53
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BTIUBNCB0053-500
- BTIUBNCB0053-100
- Antigen symbol:
- CGA
- CGA
- Antigen name:
- Glycoprotein hormones, alpha polypeptide
- Glycoprotein hormones, alpha polypeptide
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone HCGa/53
- Clone HCGa/53
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 119689
- 119689
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide