Order Entry
Export
ContactUsLinkComponent
Anti-CGA Mouse Monoclonal Antibody (Purified) [clone: HCGa/53]
Anti-CGA Mouse Monoclonal Antibody (Purified) [clone: HCGa/53]
Supplier:  Biotium
Certificates
Anti-CGA Mouse Monoclonal Antibody (Purified) [clone: HCGa/53]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BTIUBNUB0053-500
  • BTIUBNUB0053-100
  • Antigen symbol:
  • CGA
  • CGA
  • Antigen name:
  • Glycoprotein hormones, alpha polypeptide
  • Glycoprotein hormones, alpha polypeptide
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone HCGa/53
  • Clone HCGa/53
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 119689
  • 119689
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications