Order Entry
Export
ContactUsLinkComponent
Anti-TG Mouse Monoclonal Antibody (CF640R) [clone: 6E1.]
Anti-TG Mouse Monoclonal Antibody (CF640R) [clone: 6E1.]
Supplier:  Biotium
Certificates
Anti-TG Mouse Monoclonal Antibody (CF640R) [clone: 6E1.]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400051-100
  • BNC400051-500
  • Antigen symbol:
  • Tg
  • Tg
  • Antigen name:
  • Thyroglobulin
  • Thyroglobulin
  • Antigen synonyms:
  • TDH3|hTG|Tg|Tgn|AITD3
  • TDH3|hTG|Tg|Tgn|AITD3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone 6E1.
  • Clone 6E1.
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 654591
  • 654591
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications