0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Amphotericin B
Amphotericin B (from Streptomyces species) for biochemistry
Amphotericin B (from Streptomyces species) for biochemistry
Amphotericin B (from Streptomyces species) for biochemistry
Supplier:
MP Biomedicals
Certificates
Expand image
Amphotericin B (from Streptomyces species) for biochemistry
Supplier:
MP Biomedicals
Certificates
About this item
CAS Number:
1397-89-3
:
Formula:
C₄₇H₇₃NO₁₇
MDL Number:
MFCD00877763
:
924.08 g/mol
Related Information
Documentation
Certificates
Search by Lot Number
Product specification
ICNA091672348_TDS_EN
Documentation