Anti-Caspase-10 Rabbit Polyclonal Antibody
Catalog # ENZOADIAAP110E
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:Caspase 10
- Antigen symbol:CASP10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:Caspase 10 apoptosis-related cysteine peptidase|FLICE2|ALPS2|MCH4
- Amino acid number:505 to 521
- Storage buffer:PBS containing 0,02% sodium azide
- Storage temperature:4 °C
- Shipping temperature:Blue ice not frozen
- Immunogen:Synthetic peptide corresponding to aa 505-521 of human Caspase-10
- Purification:Purified by DEAE
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Caspases are synthesized as inactive zymogens that can be cleaved to form active enzymes following the induction of apoptosis by stress or death receptors.
- Dilution Range: 2 µg/ml, ECL for Western blot
- Detects a band of ~59 kDa by Western blot
Initiator caspases are activated by dimerization of the zymogen on a dedicated adaptor protein. These activated initiator caspases in-turn cleave downstream effector or executioner caspases in a cascade-like manner, which cleave key cellular proteins that lead to the morphological changes associated with apoptotic cell death.
Type: Primary
Antigen: Caspase 10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human