Order Entry
Export
ContactUsLinkComponent
 
Anti-HMOX1 Rabbit Polyclonal Antibody
 
undefined
Anti-HMOX1 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • ENZOADISPA895F
  • ENZOADISPA895D
  • Antigen symbol:
  • HMOX1
  • HMOX1
  • Antigen name:
  • Heme Oxidase 1
  • Heme Oxidase 1
  • Antigen synonyms:
  • Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
  • Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Rabbit, Sheep, Human, Dog, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant rat HO-1 (Hsp32) lacking the membrane spanning region
  • Recombinant rat HO-1 (Hsp32) lacking the membrane spanning region
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Purified by Protein A affinity chromatography
  • Purified by Protein A affinity chromatography
  • Storage buffer:
  • PBS containing 50% glycerol and 0,09% sodium azide
  • PBS containing 50% glycerol and 0,09% sodium azide
  • Storage temperature:
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice