Anti-HDJ1 Mouse Monoclonal Antibody [clone: 20]
Catalog # ENZOADISPA450E
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen symbol:HDJ1
- Clonality:Monoclonal
- Clone:2E1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Form:Liquid
- Storage buffer:PBS, pH 7,2, containing 50% glycerol
- Storage temperature:−20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human Hsp40 (Hdj1)
- Purification:Purified by Protein A affinity chromatography
- Pk:100 µG
Specifications
About this item
Hsp40/Hdj1 is a cytosolic co-chaperone belonging to the class DnaJ, named after its homolog in E. coli. Hsp40 functions in protein folding by binding nascent peptides and unfolded substrates and facilitating substrate interaction with Hsp70.
- Dilution range: Immunoprecipitation (2 - 5 µg/200 µl from 5×106cells), Western blot (1 - 2 µg/ml, ECL)
- Detects a band of ~40kDa by western blot
Type: Primary
Antigen: HDJ1
Clonality: Monoclonal
Clone: 20
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat