Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Alamethicin
Alamethicin 50%, powder
Alamethicin 50%, powder
Alamethicin 50%, powder
ICNA0215900905
:
MP Biomedicals
:
Alamethicin 50%, powder
ICNA0215900905
:
MP Biomedicals
:
Pk:
5 mg
Seq:
2210
:
:
1964.40 g/mol
:
59588-86-2
:
27061-78-5
:
C₉₂H₁₅₀N₂₂O₂₅
:
MFCD00151517