Cellulose 97.0-102.0%, microcrystalline powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9004-34-6
:
Formula:(C₁₂H₂₂O₁₁)n
MDL Number:MFCD00081512
:342.3 g/mol
Boiling Pt:Bulk Density: 0,6 g/ml (25 °C) (Lit,)
Density:0.6 g/ml (25 °C) (lit.)
Specification Test Results
- Identity Test:Passes Test
- Purity:97,0 - 102,0%
- Moisture content:<5,0%
- Particle Size +60 mesh (wt%):<8%
- Particle Size +200 mesh (wt%):>45%
- pH:5 - 7
- Residue on ignition:<0,05%
- Water Soluble Substances:<12,0 mg/5 g
- Heavy Metals:10 ppm