

N-Lauroylsarcosine sodium salt ≥94%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:137-16-6
:
Formula:H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MDL Number:MFCD00042728
:293.38 g/mol
UN:2811
ADR:6.1,II
:GHS05
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥94%
- pH (10% aqueous solution):7,5 - 8,5
- Aggregation Number:2
- CMC:14,57 mM at 30 °C (Lit.)
- Extinction Coefficient:EM = 3 (280 nm) (Lit.)