Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium iodide ≥99.0%, white solid ACS

Potassium iodide ≥99.0%, white solid ACS

100 g
Catalog # ICNA0215256280
Irritant
Supplier:   MP Biomedicals
Potassium iodide ≥99.0%, white solid ACS
Catalog # ICNA0215256280
Supplier:  MP Biomedicals
CAS Number:  7681-11-0
Potassium iodide ≥99.0%, white solid ACS

100 g

CAS Number:  7681-11-0
Price
53,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Potassium Iodide is known as Potassium Salt of Hydriodic Acid [HI]. Its Ionic form is K+ I-, and iodide ion being mild reducing agent, it can be easily oxidised to iodine by powerful oxidising agent like chlorine.

Potassium iodide is often used as a fluorescence quencher. The use of KI to facilitate the sorption of microbial proteins into dextran hydrogels from cell lysates has been described. KI has been applied in the elucidation of the crystal structure of the S100-A3 apoprotein. The use of KI in the isolation of single stranded DNA has been described.

Synonyms:  Dipotassium diiodide, Kali iodide
  • CAS Number:7681-11-0
  • Formula:Kl
  • MDL Number:MFCD00011405
  • Molecular Weight:166 g/mol
  • Melting Pt:681 °C (lit.)
  • Density:1.32 g/mL (25 °C) (lit.)
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Assay:
    ≥99,0%
  • pH (5% aq soln):
    6 - 9,2
  • Insoluble Matter:
    ≤0,005%
  • Loss on Drying (105°C):
    ≤0,2%
  • Chloride and Bromide:
    ≤0,01%
  • Iodate (IO3):
    ≤3 ppm
  • Phosphate (PO4):
    ≤0,001%
  • Sulphate (SO4):
    ≤0,005%
  • Barium (Ba):
    ≤0,002%
  • Heavy Metals (as Pb):
    ≤5 ppm
  • Iron (Fe):
    ≤3 ppm
  • Calcium (Ca):
    ≤0,002%
  • Magnesium (Mg):
    ≤0,001%
  • Sodium (Na):
    ≤0,005%

Specifications

  • Pk:
    100 g

Specifications