Order Entry
Export
ContactUsLinkComponent
Dodecamolybdophosphoric acid hydrate, powder ACS
Oxidizing
Corrosive
Dodecamolybdophosphoric acid hydrate, powder ACS
Catalog # ICNA0215255605
Supplier:  MP Biomedicals
CAS Number:  
Oxidizing
Corrosive
Dodecamolybdophosphoric acid hydrate, powder ACS
Catalog # ICNA0215255605
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Insoluble Matter:
    ≤0.01%
  • Chloride (Cl):
    <0,02%
  • Sulphate (SO4):
    <0,025%
  • Ammonium (NH4):
    0,01%
  • Calcium (Ca):
    0,02%
  • Heavy Metals (as Pb):
    <0,005%
  • Iron (Fe) :
    <0,005%

Specification Test Results

About this item

 :
 :1843.27 g/mol
Storage Temperature:Ambient
UN:3260
ADR:8,III
 :GHS03
 :GHS05
CAS Number:51429-74-4
Formula:H₃[P(Mo₃O₁₀)₄]·nH₂O
MDL Number:MFCD00149913