0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Caffeine
Caffeine, anhydrous, powder
Caffeine, anhydrous, powder
Caffeine, anhydrous, powder
Supplier:
MP Biomedicals
Certificates
Expand image
Caffeine, anhydrous, powder
Supplier:
MP Biomedicals
Certificates
About this item
CAS Number:
58-08-2
:
Formula:
C₈H₁₀N₄O₂
MDL Number:
MFCD00005758
:
194.19 g/mol
Related Information
Documentation
Certificates
Search by Lot Number
Documentation