D-(+)-Trehalose dihydrate ≥98%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:6138-23-4
:
Formula:C₁₂H₂₂O₁₁·2H₂O
MDL Number:MFCD00071594
:378.33 g/mol
Melting Pt:203 °C (lit.)
Storage Temperature:Ambient
EINECS:202-739-6
Specification Test Results
- Identity Test:Passes
- Purity:≥98%
- FTIR:Conforms to Standard
- Loss on Drying:8.5 - 10.5%