Order Entry
Export
ContactUsLinkComponent
 
L(-)-Proline ≥99%, white powder
 
L(-)-Proline ≥99%, white powder

 

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • Loss on Drying:
    ≤0.4%
  • Optical Rotation (c=1.6, water):
    -82° to -85°
  • Hydroxy-L-proline:
    Free