D-(+)-Melibiose monohydrate, white powder
:66009-10-7
:585-99-9
:C₁₂H₂₂O₁₁·H₂O
:MFCD00198188
:360.312 g/mol
- Identity Test:Passes Test
- FTIR:Conforms to Standard
- Melting Point:174 - 190°C
- Moisture Content:≤5%
- Optical Rotation (c=4, water):+135 ± 10°