
2,2'-Bipyridyl ≥99.5%, powder
ICNA0210161425
:
- Pk:25 g
- Seq:3308
- Identity Test:Passes
- Purity:≥99.5%
- FTIR:Conforms to Standard
- Melting Point:65 - 75 °C
:
:156.19 g/mol
:72 °C
:Ambient
:2811
:6.1,III
:GHS06
:366-18-7
:C₁₀H₈N₂
:MFCD00006212