Order Entry
Export
ContactUsLinkComponent
Human THBS1 ELISA kit
Human THBS1 ELISA kit
Catalog # BIRBORB96799-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human THBS1 ELISA kit
Catalog # BIRBORB96799-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Primary antibody reactivity:
    Human
  • Description:
    Human THBS1 ELISA Kit
  • Specificity:
    Natural and recombinant human THBS1
  • Detection range:
    1,56 - 100 ng/ml
  • Sensitivity:
    <50 pg/ml

Specifications

About this item

Human THBS1 ELISA Kit is a sandwich ELISA kit for the quantitative detection of human Thrombospondin 1.

  • No detectable cross-reactivity with other relevant proteins

Thrombospondin 1, also known as THBS1, is a protein that in humans in encoded by the THBS1 gene. Thrombospondin 1 is a subunit of a disulfide-linked homotrimeric protein. This protein is an adhesive glycoprotein that mediates cell-to-cell and cell-to-matrix interactions. By in situ hybridization, the THBS1 gene was mapped to human 15q15 and the cognate gene to mouse chromosome 2 region F. In 1990, the THBS1 gene was localized to 15q11-qter by Southern analysis of human-rodent somatic cell hybrids. Thrombospondin I is a multimodular secreted protein that associates with the extracellular matrix and possesses a variety of biologic functions, including a potent antiangiogenic activity. Other thrombospondin genes include thrombospondins II, III, and IV.