


Sodium dodecyl sulphate (SDS) ≥99%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
:288.38 g/mol
Melting Pt:180…210 °C
Density:>1.1 (20 °C)
Flash Pt:199 °C
EINECS:205-788-1
UN:2296
ADR:4.1,II
:GHS02
:GHS06
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:~99%
- FTIR:Conforms to Standard
- Melting Point:182 - 210 °C
- OD260 nm (1% aq soln):<0,1
- OD280 nm (1% aq soln) :<0,1
- pH (1% aq soln):6 - 7