Order Entry
Export
ContactUsLinkComponent
L(-)-Glutathione (oxidised form), crystalline powder
L(-)-Glutathione (oxidised form), crystalline powder
Supplier:  MP Biomedicals
Certificates
L(-)-Glutathione (oxidised form), crystalline powder
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • pH (3.3% aq soln):
    Acidic to Litmus Paper
  • Solubility (3.3% aq soln):
    Clear, Colorless Solution
  • Optical Rotation (c=1, water):
    –85 ° to –110 °
  • FTIR:
    Conforms to Standard
  • Moisture:
    For Information Only

Specification Test Results