
Methyl-4-hydroxybenzoate, white crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:99-76-3
:
Formula:HOC₆H₄CO₂CH₃
MDL Number:MFCD00002352
:152.15 g/mol
Boiling Pt:270…280 °C
Melting Pt:124…131 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- Melting Point:124 - 131 °C
- UV/Vis: l max (methanol):255 ± 3 nm
- Solubility:Passes
- Acidity:Passes
- Residue on Ignition:≤0.1%