ESD safety shoes, Velcro® fastening, SL 465 XP Blue
Lightweight ESD safety shoe made from Sportline upper (oiled nubuck leather), breathable functional lining and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
- Moisture control and breathability thanks to Clima-stream® technology
- Removable insoles; orthopedic insoles possible
- 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
- Puncture resistant, metal-free XP® midsole
- Lightweight alu-tec® toecap made from aluminium (200 J)
- Carbon fibre reinforced toe protection
- Electrical contact resistance of 1×10⁵ to 3,5×10⁷ Ohm
Colour: Black/blue
: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC