Anti-NFIA Rabbit Polyclonal Antibody
BSBTPB10080
- Antibody type:Primary
- Antigen name:Nuclear factor I/A
- Antigen symbol:NFIA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4774
- Antigen synonyms:NFI-A|NF1-A|CTF|NFI-L|NF-I/A
- Size:100 μg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Nuclear factor 1 A-type(NFIA) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: NFIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat