Order Entry
Export
ContactUsLinkComponent
 
Anti-APOB Rabbit Polyclonal Antibody
  BSBTPB10042
undefined
Anti-APOB Rabbit Polyclonal Antibody
  BSBTPB10042

 

  • Antibody type:
    Primary
  • Antigen name:
    Apolipoprotein B
  • Antigen symbol:
    APOB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    338
  • Antigen synonyms:
    apoB-100|FLDB|apoB-48|LDLCQ4
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Apolipoprotein B-100(APOB) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: APOB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human