Anti-PanCK Mouse Monoclonal Antibody (Fluorescein isothiocyanate (FITC)) [clone: 3K]
Catalog # ABNODH0025
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:PanCK
- Clonality:Monoclonal
- Clone:3K
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:20 reactions
- Pk:20 Assays
Specifications
About this item
Anti-PanCK monoclonal antibody (FITC) is a FITC-conjugated monoclonal antibody for identification of cells expressing the pan cytokeratin antigen.
Type: Primary
Antigen: PanCK
Clonality: Monoclonal
Clone: 3K
Conjugation: Fluorescein isothiocyanate (FITC)
Epitope:
Host: Mouse
Isotype: IgG1, Kappa
Reactivity: Human