Anti-CD19 Mouse Monoclonal Antibody (APC) [clone: A3-B1]
Catalog # ABNOMAB13831
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:CD19
- Clonality:Monoclonal
- Clone:A3-B1
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:930
- Size:100 reactions
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human CD19.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: A3-B1
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human