Order Entry
Export
ContactUsLinkComponent
Anti-ITGAL Mouse Monoclonal Antibody (PerCP) [clone: TP1/40]
Anti-ITGAL Mouse Monoclonal Antibody (PerCP) [clone: TP1/40]
Catalog # ABNOMAB13787
Supplier:  Abnova
Anti-ITGAL Mouse Monoclonal Antibody (PerCP) [clone: TP1/40]
Catalog # ABNOMAB13787
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ITGAL
  • Clonality:
    Monoclonal
  • Clone:
    TP1/40
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Gene ID:
    3683
  • Size:
    100 reactions
  • Pk:
    100 Assays

Specifications

About this item

Mouse monoclonal antibody raised against human ITGAL.

Type: Primary
Antigen: ITGAL
Clonality: Monoclonal
Clone: TP1/40
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human