Anti-SPN6 Mouse Monoclonal Antibody (PerCP) [clone: TP1/36]
ABNOMAB13918
: Abnova
- Antibody type:Primary
- Antigen symbol:SPN6
- Clonality:Monoclonal
- Clone:TP1/36
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:6693
- Size:100 reactions
- Pk:100 Assays
Mouse monoclonal antibody raised against native human SPN.
Type: Primary
Antigen: SPN6
Clonality: Monoclonal
Clone: TP1/36
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human