Order Entry
Export
ContactUsLinkComponent
Anti-SPN6 Mouse Monoclonal Antibody (FITC) [clone: TP1/36]
Anti-SPN6 Mouse Monoclonal Antibody (FITC) [clone: TP1/36]
Catalog # ABNOMAB13915
Supplier:  Abnova
Anti-SPN6 Mouse Monoclonal Antibody (FITC) [clone: TP1/36]
Catalog # ABNOMAB13915
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SPN6
  • Clonality:
    Monoclonal
  • Clone:
    TP1/36
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Gene ID:
    6693
  • Size:
    100 reactions
  • Pk:
    100 Assays

Specifications

About this item

Mouse monoclonal antibody raised against native human SPN.

Type: Primary
Antigen: SPN6
Clonality: Monoclonal
Clone: TP1/36
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human