Anti-CD21 Mouse Monoclonal Antibody (APC) [clone: TP1/31]
ABNOMAB13673
: Abnova
- Antibody type:Primary
- Antigen symbol:CD21
- Clonality:Monoclonal
- Clone:TP1/31
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:914
- Size:100 reactions
- Pk:100 Assays
Mouse monoclonal antibody raised against human CD2.
Type: Primary
Antigen: CD21
Clonality: Monoclonal
Clone: TP1/31
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human