Anti-ITGAL0 Mouse Monoclonal Antibody (FITC) [clone: TP1/40]
Catalog # ABNOMAB13784
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:ITGAL0
- Clonality:Monoclonal
- Clone:TP1/40
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:3683
- Size:100 reactions
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human ITGAL.
Type: Primary
Antigen: ITGAL0
Clonality: Monoclonal
Clone: TP1/40
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human