Anti-CD3E3 Mouse Monoclonal Antibody (CF-Blue) [clone: 33-2A3]
ABNOMAB13723
: Abnova
- Antibody type:Primary
- Antigen symbol:CD3E3
- Clonality:Monoclonal
- Clone:33-2A3
- Conjugation:CF-Blue™
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Gene ID:916
- Size:100 reactions
- Pk:100 Assays
Mouse monoclonal antibody raised against human CD3E.
Type: Primary
Antigen: CD3E3
Clonality: Monoclonal
Clone: 33-2A3
Conjugation: CF-Blue
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human