Anti-CD53 Mouse Monoclonal Antibody (FITC) [clone: JS11]
ABNOMAB13712
: Abnova
- Antibody type:Primary
- Antigen symbol:CD53
- Clonality:Monoclonal
- Clone:JS11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Epitope:Full-length
- Gene ID:1604
- Size:100 reactions
- Pk:100 Assays
Mouse monoclonal antibody raised against full length recombinant human CD55.
Type: Primary
Antigen: CD53
Clonality: Monoclonal
Clone: JS11
Conjugation: FITC
Epitope: Full-length
Host: Mouse
Isotype: IgG1
Reactivity: Human