Anti-CD53 Mouse Monoclonal Antibody (PE) [clone: HI29]
Catalog # ABNOMAB13707
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:CD53
- Clonality:Monoclonal
- Clone:HI29
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:963
- Size:100 reactions
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human CD53.
Type: Primary
Antigen: CD53
Clonality: Monoclonal
Clone: HI29
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human