Anti-CD52 Mouse Monoclonal Antibody (APC) [clone: L17F12]
Catalog # ABNOMAB13742
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:CD52
- Clonality:Monoclonal
- Clone:L17F12
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Gene ID:921
- Size:100 reactions
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human CD5.
Type: Primary
Antigen: CD52
Clonality: Monoclonal
Clone: L17F12
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human