Order Entry
Export
ContactUsLinkComponent
Anti-CYCSC12 Mouse Monoclonal Antibody [clone: 7H8.2C12]
Anti-CYCSC12 Mouse Monoclonal Antibody [clone: 7H8.2C12]
Catalog # ABNOMAB13397
Supplier:  Abnova
Anti-CYCSC12 Mouse Monoclonal Antibody [clone: 7H8.2C12]
Catalog # ABNOMAB13397
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CYCSC12
  • Clonality:
    Monoclonal
  • Clone:
    7H8.2C12
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Pigeon,
    Human
  • Gene ID:
    54205
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against synthetic peptide of CYCS.

Type: Primary
Antigen: CYCSC12
Clonality: Monoclonal
Clone: 7H8.2C12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human,Pigeon