Anti-IPO-388 Mouse Monoclonal Antibody [clone: IPO-38]
Catalog # ABNOMAB13232
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:IPO-388
- Clonality:Monoclonal
- Clone:IPO-38
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against native human IPO-38.
Type: Primary
Antigen: IPO-388
Clonality: Monoclonal
Clone: IPO-38
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human