Anti-Blood Group Antigen H Type 2E Mouse Monoclonal Antibody [clone: 19-OLE]
Catalog # ABNOMAB13291
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Blood Group Antigen H Type 2E
- Antigen symbol:Blood Group Antigen H Type 2E
- Clonality:Monoclonal
- Clone:19-OLE
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against native human Blood Group Antigen H Type 2.
Type: Primary
Antigen: Blood Group Antigen H Type 2E
Clonality: Monoclonal
Clone: 19-OLE
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human