Anti-Streptococcus group A Rabbit Polyclonal Antibody
500 μg
Catalog # ABNOPAB29816
Supplier: Abnova
Jump to:
Product Details & Documents
Rabbit polyclonal antibody raised against Streptococcus group A.
Type: Primary
Antigen: Streptococcus group A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Streptococcus
Specifications
- Size:500 μg
- Cat. No.:ABNOPAB29816
- Clonality:Polyclonal
- Reactivity:Streptococcus
- Antibody type:Primary
- Host:Rabbit
- Antigen symbol:Streptococcus group A
- Antigen name:Streptococcus group A antigen



