Anti-Streptococcus group A Rabbit Monoclonal Antibody [clone: HM133]
Catalog # ABNOMAB13654
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Streptococcus group A
- Clonality:Monoclonal
- Clone:HM133
- Host:Rabbit
- Isotype:IgG1
- Reactivity:Streptococcus
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Rabbit monoclonal antibody raised against Streptococcus group A.
Type: Primary
Antigen: Streptococcus group A
Clonality: Monoclonal
Clone: HM133
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Streptococcus