Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-FOXF2 Rabbit Polyclonal Antibody

Anti-FOXF2 Rabbit Polyclonal Antibody

100 μL
Catalog # ABNOPAB30084
Supplier:   Abnova
Anti-FOXF2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB30084
Supplier:  Abnova
Anti-FOXF2 Rabbit Polyclonal Antibody
Anti-FOXF2 Rabbit Polyclonal Antibody

100 μL

Price
523,00 €
Log in for availability
1 * 100 µl
Product Details & Documents

Rabbit polyclonal antibody raised against synthetic peptide of human FOXF2.

Type: Primary
Antigen: FOXF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Specifications
  • Size:
    100 μL
  • Western blot:
    Yes
  • Cat. No.:
    ABNOPAB30084
  • Clonality:
    Polyclonal
  • Reactivity:
    Human
  • Antibody type:
    Primary
  • Gene ID:
    2295
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Sequence:
    SSASCASSSSSSNSASAPSAACKSAGGGGAGAGSGGAKKASSGLRRPEKP
  • Antigen symbol:
    FOXF2