Anti-GABRP Rabbit Polyclonal Antibody
Catalog # ABNOPAB30092
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:GABRP
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:2568
- Sequence:VEVGRSDKLSLPGFENLTAGYNKFLRPNFGGEPVQIALTLDIASISSISE
- Size:100 μL
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of human GABRP.
Type: Primary
Antigen: GABRP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human