BrightVision+ Anti-IgG (Poly-HRP)
Catalog # DPVR110HRP
Supplier: IMMUNOLOGIC
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Conjugation:Poly-HRP (Horseradish Peroxidase)
- ImmunoChemistry:Yes
- Reactivity:Rabbit
- Seq:03
- Storage temperature:+2 °C to +8 °C
- Size:110 mL
- Pk:110 mL
Specifications
About this item
BrightVision histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric HRP linker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, this kit has the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies.
- High-quality secondary antibody for IHC applications, HRP labelled
- Antigen specificity: Anti-Rabbit IgG (H+L)
Biotin-free
Ready-to-use
Certifications: Meets CE/IVD standards.
Ordering information: Packages above 110 ml contain small empty bottles so users can create smaller working volumes and prevent possible contamination of the whole lot.
Delivery: Shipped with blue ice. Store between 2 to 8 °C.
Caution: Irritating to eyes, respiratory systems and skin.