BrightVision+ Anti-IgG Antibody (Poly-HRP)
Supplier: IMMUNOLOGIC
Certificates
About this item
BrightVision histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric AP linker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, BrightVision+histostaining kits have the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies.
- High-quality secondary antibody for IHC applications, HRP labelled
- Antigen Specificity: Anti-mouse IgG (H+L)
Biotin-free
Ready-to-use
Certifications: Meets CE/IVD standards.
Ordering information: Packages above 110 ml contain small empty bottles so users can create smaller working volumes and prevent possible contamination of the whole lot.
Delivery: Shipped with blue ice. Store between 2 to 8 °C.
Caution: Irritating to eyes, respiratory systems and skin.
Specifications
- Catalog No:
- DPVM55HRP
- DPVM110HRP
- Antigen symbol:
- IgG (H+L)
- IgG (H+L)
- Antibody type:
- Secondary
- Secondary
- Conjugation:
- Poly-HRP (Horseradish Peroxidase)
- Poly-HRP (Horseradish Peroxidase)
- Reactivity:
- Mouse
- Mouse
- ImmunoChemistry:
- Yes
- Yes
- Storage temperature:
- +2 °C to +8 °C
- +2 °C to +8 °C