Order Entry
Export
ContactUsLinkComponent
BrightVision+ Anti-IgG (Poly-AP)
BrightVision+ Anti-IgG (Poly-AP)
Catalog # DPVM55AP
Supplier:  IMMUNOLOGIC
BrightVision+ Anti-IgG (Poly-AP)
Catalog # DPVM55AP
Supplier:  IMMUNOLOGIC

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Conjugation:
    Poly-AP (Alkaline Phosphatase)
  • Reactivity:
    Mouse
  • Seq:
    02
  • Storage temperature:
    +2 °C to +8 °C
  • Pk:
    55 mL

Specifications

About this item

BrightVision histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric AP linker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, BrightVision histostaining kits have the advantages of higher amplification power, biotin-free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies.

  • Antigen Specificity: Anti-Mouse IgG (H+L)
  • High quality secondary antibody for IHC, AP labelled

Biotin-free
Ready-to-use

Certifications: Meets CE/IVD standards.
Ordering information: Packages above 110 ml contain small empty bottles so users can create smaller working volumes and prevent possible contamination of the whole lot.
Delivery: The Immunologic products will be shipped with blue ice. Store the products between 2 to 8 °C.
Caution: Irritating to eyes, respiratory systems and skin.