Order Entry
Export
ContactUsLinkComponent
 
BrightVision+ Anti-IgG (Poly-AP)
  DPVB110AP
BrightVision+ Anti-IgG (Poly-AP)
  DPVB110AP

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Conjugation:
    Poly-AP (Alkaline Phosphatase)
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Rabbit,
    Mouse
  • Seq:
    03
  • Storage temperature:
    +2 °C to +8 °C
  • Pk:
    110 mL

 

 

BrightVision+ histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric APlinker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, BrightVision+ histostaining kits have the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies.

  • High quality secondary antibody for IHC applications, AP labelled
  • Antigen specificity: Anti-Mouse IgG (H+L), Anti-Rabbit IgG (H+L)
  • Kit contains post-antibody blocking (ready-to-use) and poly-AP-anti-mouse/rabbit IgG (ready-to-use)

Colour version available (see IMMS/C-DPVB-AP and similar articles).

Biotin-free
Ready-to-use

 : Meets CE/IVD standards.
 : Packages above 110 ml contain small empty bottles so users can create smaller working volumes and prevent possible contamination of the whole lot.
 : Shipped with blue ice. Store between 2 to 8 °C.
 : Irritating to eyes, respiratory systems and skin.