Order Entry
Export
ContactUsLinkComponent
Anti-CD25 Rat Monoclonal Antibody (APC-Cyanine7) [clone: PC61.5]
Anti-CD25 Rat Monoclonal Antibody (APC-Cyanine7) [clone: PC61.5]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD25 Rat Monoclonal Antibody (APC-Cyanine7) [clone: PC61.5]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 25-0251-U100
  • 25-0251-U025
  • Antigen symbol:
  • CD25
  • CD25
  • Antigen name:
  • CD25, Interleukin-2 Receptor alpha chain
  • CD25, Interleukin-2 Receptor alpha chain
  • Antigen synonyms:
  • Interleukin-2 Receptor alpha|Tac|p55|IL-2R alpha|Ly-43
  • Interleukin-2 Receptor alpha|Tac|p55|IL-2R alpha|Ly-43
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)-Cy7®
  • APC (Allophycocyanin)-Cy7®
  • Clone:
  • PC61.5
  • PC61.5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16184
  • 16184
  • Isotype:
  • IgG lambda
  • IgG lambda
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • +2 °C to +8 °C, protected from light
  • +2 °C to +8 °C, protected from light

Specifications

Related Information